Product Details

SNP ID
rs3783608
Assay Type
Functionally tested
NCBI dbSNP Submissions
35
Location
Chr.1:100719752 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCTATTTCACTCCGCGGTATCTG[C/T]ATCGGGCCTCACTGGCTTCAGGAGC
Phenotype
MIM: 192225
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VCAM1 PubMed Links

Gene Details

Gene
VCAM1
Gene Name
vascular cell adhesion molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001078.3 113 UTR 5 NP_001069.1
NM_001199834.1 113 UTR 5 NP_001186763.1
NM_080682.2 113 UTR 5 NP_542413.1

View Full Product Details