Product Details

SNP ID
rs3730192
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:102398536 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGCTCTTGATCAAGTTCAGAAGA[C/T]TGGATAATATATTGCTAATTTTGCT
Phenotype
MIM: 147440
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGF1 PubMed Links

Gene Details

Gene
IGF1
Gene Name
insulin like growth factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000618.4 4499 UTR 3 NP_000609.1
NM_001111283.2 4499 UTR 3 NP_001104753.1
NM_001111284.1 4499 UTR 3 NP_001104754.1
XM_017019259.1 4499 Intron XP_016874748.1
XM_017019260.1 4499 Intron XP_016874749.1
XM_017019261.1 4499 Intron XP_016874750.1
XM_017019262.1 4499 UTR 3 XP_016874751.1
XM_017019263.1 4499 UTR 3 XP_016874752.1

View Full Product Details