Product Details

SNP ID
rs3814514
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33510878 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTCCTCGTAAGAGGATCTCCCC[A/G]GTTTCTCGCGGTCCTGCTAAAACGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SUGT1P1 PubMed Links

Gene Details

Gene
SUGT1P1
Gene Name
SGT1 homolog, MIS12 kinetochore complex assembly cochaperone pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details