Product Details

SNP ID
rs3736582
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:122983379 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCTCCTGAGACCATCCACCTGATG[C/G]GAAGAAAGCTAGAAAAGCCCAACCC
Phenotype
MIM: 606238 MIM: 611310
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IKZF5 PubMed Links

Gene Details

Gene
IKZF5
Gene Name
IKAROS family zinc finger 5
There are no transcripts associated with this gene.

Gene
PSTK
Gene Name
phosphoseryl-tRNA kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153336.2 682 Missense Mutation CGA,GGA R206G NP_699167.2
XM_017015640.1 682 Missense Mutation CGA,GGA R206G XP_016871129.1
XM_017015641.1 682 Missense Mutation CGA,GGA R206G XP_016871130.1

View Full Product Details