Product Details

SNP ID
rs3738512
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:42813477 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TCAAAGGAACTTCTGGGCTCTTACT[C/G]TTAGTGACATCTAATCTGGATGGTT
Phenotype
MIM: 609017
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ERMAP PubMed Links

Gene Details

Gene
ERMAP
Gene Name
erythroblast membrane associated protein (Scianna blood group)
There are no transcripts associated with this gene.

Gene
SVBP
Gene Name
small vasohibin binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199342.3 Intron NP_955374.1
XM_017001226.1 Intron XP_016856715.1

View Full Product Details