Product Details

SNP ID
rs3740584
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:117485222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGCTGTGGACAGCATAGAGTGG[A/G]GTGGGCGGAAAAGGGTTGGTGCAAG
Phenotype
MIM: 607637
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EMX2OS PubMed Links

Gene Details

Gene
EMX2OS
Gene Name
EMX2 opposite strand/antisense RNA
There are no transcripts associated with this gene.

View Full Product Details