Product Details

SNP ID
rs3807551
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:50592707 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATTCCAGTTTATTTTCAACTTTCC[A/G]CTGGATCTTCCATGCCCTCCCCAAT
Phenotype
MIM: 601523
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRB10 PubMed Links

Gene Details

Gene
GRB10
Gene Name
growth factor receptor bound protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001549.2 2333 UTR 3 NP_001001549.1
NM_001001550.2 2333 UTR 3 NP_001001550.1
NM_001001555.2 2333 UTR 3 NP_001001555.1
NM_005311.4 2333 UTR 3 NP_005302.3
XM_011515304.1 2333 UTR 3 XP_011513606.1
XM_011515312.2 2333 UTR 3 XP_011513614.1
XM_011515316.1 2333 UTR 3 XP_011513618.1
XM_011515323.2 2333 UTR 3 XP_011513625.1
XM_017012029.1 2333 UTR 3 XP_016867518.1
XM_017012030.1 2333 UTR 3 XP_016867519.1
XM_017012031.1 2333 UTR 3 XP_016867520.1
XM_017012032.1 2333 UTR 3 XP_016867521.1
XM_017012033.1 2333 UTR 3 XP_016867522.1
XM_017012034.1 2333 UTR 3 XP_016867523.1
XM_017012035.1 2333 UTR 3 XP_016867524.1
XM_017012036.1 2333 UTR 3 XP_016867525.1
XM_017012037.1 2333 UTR 3 XP_016867526.1
XM_017012038.1 2333 UTR 3 XP_016867527.1
XM_017012039.1 2333 UTR 3 XP_016867528.1
XM_017012040.1 2333 UTR 3 XP_016867529.1
XM_017012041.1 2333 UTR 3 XP_016867530.1
XM_017012042.1 2333 UTR 3 XP_016867531.1
XM_017012043.1 2333 UTR 3 XP_016867532.1
XM_017012044.1 2333 UTR 3 XP_016867533.1
XM_017012045.1 2333 UTR 3 XP_016867534.1
XM_017012046.1 2333 UTR 3 XP_016867535.1
XM_017012047.1 2333 UTR 3 XP_016867536.1
XM_017012048.1 2333 UTR 3 XP_016867537.1
XM_017012049.1 2333 UTR 3 XP_016867538.1
XM_017012050.1 2333 UTR 3 XP_016867539.1
XM_017012051.1 2333 UTR 3 XP_016867540.1
XM_017012052.1 2333 UTR 3 XP_016867541.1
XM_017012053.1 2333 UTR 3 XP_016867542.1
XM_017012054.1 2333 UTR 3 XP_016867543.1
XM_017012055.1 2333 UTR 3 XP_016867544.1
XM_017012056.1 2333 UTR 3 XP_016867545.1
XM_017012057.1 2333 UTR 3 XP_016867546.1
XM_017012058.1 2333 UTR 3 XP_016867547.1
XM_017012059.1 2333 UTR 3 XP_016867548.1
XM_017012060.1 2333 UTR 3 XP_016867549.1
XM_017012061.1 2333 UTR 3 XP_016867550.1
XM_017012062.1 2333 UTR 3 XP_016867551.1
XM_017012063.1 2333 UTR 3 XP_016867552.1
XM_017012064.1 2333 UTR 3 XP_016867553.1
XM_017012065.1 2333 UTR 3 XP_016867554.1
XM_017012066.1 2333 UTR 3 XP_016867555.1
XM_017012067.1 2333 UTR 3 XP_016867556.1
XM_017012068.1 2333 UTR 3 XP_016867557.1

View Full Product Details