Product Details

SNP ID
rs3813082
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:49561673 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACTCCCAGAGAGAGTGTGGCTTTG[A/C]GCCTTGGAGAGGATGCTCTCCTTCT
Phenotype
MIM: 603684
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LIPG PubMed Links

Gene Details

Gene
LIPG
Gene Name
lipase G, endothelial type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308006.1 188 Intron NP_001294935.1
NM_006033.3 188 Intron NP_006024.1
XM_005258390.1 188 UTR 5 XP_005258447.1
XM_011526265.2 188 UTR 5 XP_011524567.1
XM_011526267.1 188 Intron XP_011524569.1
XM_017026095.1 188 Intron XP_016881584.1

View Full Product Details