Product Details

SNP ID
rs3825736
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:69612748 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGACATGCTGGGCTTTGTTGCTCTC[A/G]CTAACTGGGCTCCTGTGTGCTCTTC
Phenotype
MIM: 616761
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SUSD6 PubMed Links

Gene Details

Gene
SUSD6
Gene Name
sushi domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014734.3 Intron NP_055549.1
XM_011537412.1 Intron XP_011535714.1
XM_011537413.1 Intron XP_011535715.1
XM_017021838.1 Intron XP_016877327.1
XM_017021839.1 Intron XP_016877328.1

View Full Product Details