Product Details

SNP ID
rs3781239
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:92458040 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTGTCACCCAAACCTTCATTACCC[C/G]CTTGTCTACCCACATCAGCAGACAG
Phenotype
MIM: 146680
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IDE PubMed Links

Gene Details

Gene
IDE
Gene Name
insulin degrading enzyme
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165946.1 Intron NP_001159418.1
NM_001322793.1 Intron NP_001309722.1
NM_001322794.1 Intron NP_001309723.1
NM_001322795.1 Intron NP_001309724.1
NM_001322796.1 Intron NP_001309725.1
NM_001322797.1 Intron NP_001309726.1
NM_004969.3 Intron NP_004960.2
XM_017016187.1 Intron XP_016871676.1
XM_017016188.1 Intron XP_016871677.1
XM_017016189.1 Intron XP_016871678.1
XM_017016190.1 Intron XP_016871679.1

View Full Product Details