Product Details
- SNP ID
-
rs3816146
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
49
- Location
-
Chr.1:160541858 on Build GRCh38
- Set Membership
-
HapMap
JSNP
- Context Sequence [VIC/FAM]
- GATTGCTCTTTATGTAATAGCAACC[C/T]AGGTAACACGTGATAAAGCAATGAT
- Phenotype
-
MIM: 604513
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CD84
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75719052] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CD84
- Gene Name
- CD84 molecule
View Full Product Details