Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001271602.1 | Intron | NP_001258531.1 | |||
NM_001318505.1 | Intron | NP_001305434.1 | |||
NM_014606.2 | Intron | NP_055421.1 | |||
XM_005263327.3 | Intron | XP_005263384.1 | |||
XM_005263329.3 | Intron | XP_005263386.1 | |||
XM_005263330.3 | Intron | XP_005263387.1 | |||
XM_005263331.1 | Intron | XP_005263388.1 | |||
XM_005263332.3 | Intron | XP_005263389.1 | |||
XM_011532397.2 | Intron | XP_011530699.1 | |||
XM_017008806.1 | Intron | XP_016864295.1 | |||
XM_017008807.1 | Intron | XP_016864296.1 | |||
XM_017008808.1 | Intron | XP_016864297.1 |