Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_003668.3 | 8986 | Intron | NP_003659.2 | ||
NM_139078.2 | 8986 | Intron | NP_620777.1 | ||
XM_017020125.1 | 8986 | Intron | XP_016875614.1 | ||
XM_017020126.1 | 8986 | Intron | XP_016875615.1 | ||
XM_017020127.1 | 8986 | UTR 3 | XP_016875616.1 | ||
XM_017020128.1 | 8986 | UTR 3 | XP_016875617.1 | ||
XM_017020129.1 | 8986 | Intron | XP_016875618.1 | ||
XM_017020130.1 | 8986 | Intron | XP_016875619.1 | ||
XM_017020131.1 | 8986 | Intron | XP_016875620.1 | ||
XM_017020132.1 | 8986 | UTR 3 | XP_016875621.1 | ||
XM_017020133.1 | 8986 | UTR 3 | XP_016875622.1 | ||
XM_017020134.1 | 8986 | UTR 3 | XP_016875623.1 | ||
XM_017020135.1 | 8986 | UTR 3 | XP_016875624.1 | ||
XM_017020136.1 | 8986 | UTR 3 | XP_016875625.1 | ||
XM_017020137.1 | 8986 | Intron | XP_016875626.1 | ||
XM_017020138.1 | 8986 | UTR 3 | XP_016875627.1 | ||
XM_017020139.1 | 8986 | Intron | XP_016875628.1 |