Product Details

SNP ID
rs3749513
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:68929679 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGATATACTCACAACCTCATGATC[A/G]TGGAATTCTAGGCTATATAGCTAAG
Phenotype
MIM: 616382
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UGT2A3 PubMed Links

Gene Details

Gene
UGT2A3
Gene Name
UDP glucuronosyltransferase family 2 member A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024743.3 1749 UTR 3 NP_079019.3
XM_011532247.2 1749 UTR 3 XP_011530549.1

View Full Product Details