Product Details

SNP ID
rs3829956
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:18316521 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGCCAGTGTGGAGAAGTCCATCAT[C/T]GAACATCAAGACCTGCTGAAGCAGA
Phenotype
MIM: 601243
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SMCR8 PubMed Links

Gene Details

Gene
SMCR8
Gene Name
Smith-Magenis syndrome chromosome region, candidate 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144775.2 1242 Silent Mutation ATC,ATT I244I NP_658988.2
Gene
TOP3A
Gene Name
topoisomerase (DNA) III alpha
There are no transcripts associated with this gene.

View Full Product Details