Product Details
- SNP ID
-
rs4080720
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:43746695 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGCGGGCCTCATGCTCGTCGAGTT[C/T]TTCGCCCCCTGGTGAGTCCATTCTG
- Phenotype
-
MIM: 602046
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CATSPER2P1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2411284] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CATSPER2P1
- Gene Name
- cation channel sperm associated 2 pseudogene 1
There are no transcripts associated with this gene.
- Gene
- PDIA3
- Gene Name
- protein disulfide isomerase family A member 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005313.4 |
304 |
Silent Mutation |
TTC,TTT |
F52F |
NP_005304.3 |
View Full Product Details