Product Details

SNP ID
rs28933684
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:43949831 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCATTCCTCGCAGTGACAGGAGA[G/A]GATGTACCGGTAGGTGGCAGTGAGT
Phenotype
MIM: 300658
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
NDP PubMed Links

Gene Details

Gene
NDP
Gene Name
NDP, norrin cystine knot growth factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000266.3 949 Missense Mutation CTC,TTC L124F NP_000257.1

View Full Product Details