Product Details

SNP ID
rs75830605
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:47272842 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGGACAAGGACTCATCTGTGGGT[A/G]TTGTATTACTGATTCTGAGCCTGAG
Phenotype
MIM: 603584 MIM: 602423
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101928943 PubMed Links

Gene Details

Gene
LOC101928943
Gene Name
uncharacterized LOC101928943
There are no transcripts associated with this gene.

Gene
MADD
Gene Name
MAP kinase activating death domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135943.1 Intron NP_001129415.1
NM_001135944.1 Intron NP_001129416.1
NM_003682.3 Intron NP_003673.3
NM_130470.2 Intron NP_569826.2
NM_130471.2 Intron NP_569827.2
NM_130472.2 Intron NP_569828.2
NM_130473.2 Intron NP_569829.2
NM_130474.2 Intron NP_569830.2
NM_130475.2 Intron NP_569831.1
NM_130476.2 Intron NP_569832.2
XM_005253189.2 Intron XP_005253246.1
XM_005253196.2 Intron XP_005253253.1
XM_005253199.2 Intron XP_005253256.1
XM_005253200.2 Intron XP_005253257.1
XM_005253201.2 Intron XP_005253258.1
XM_005253203.2 Intron XP_005253260.1
XM_005253204.2 Intron XP_005253261.1
XM_005253205.1 Intron XP_005253262.1
XM_017018478.1 Intron XP_016873967.1
XM_017018479.1 Intron XP_016873968.1
XM_017018480.1 Intron XP_016873969.1
XM_017018481.1 Intron XP_016873970.1
XM_017018482.1 Intron XP_016873971.1
XM_017018483.1 Intron XP_016873972.1
XM_017018484.1 Intron XP_016873973.1
XM_017018485.1 Intron XP_016873974.1
XM_017018486.1 Intron XP_016873975.1
XM_017018487.1 Intron XP_016873976.1
XM_017018488.1 Intron XP_016873977.1
XM_017018489.1 Intron XP_016873978.1
XM_017018490.1 Intron XP_016873979.1
XM_017018491.1 Intron XP_016873980.1
XM_017018492.1 Intron XP_016873981.1
XM_017018493.1 Intron XP_016873982.1
XM_017018494.1 Intron XP_016873983.1
XM_017018495.1 Intron XP_016873984.1
XM_017018496.1 Intron XP_016873985.1
XM_017018497.1 Intron XP_016873986.1
XM_017018498.1 Intron XP_016873987.1
XM_017018499.1 Intron XP_016873988.1
XM_017018500.1 Intron XP_016873989.1
XM_017018501.1 Intron XP_016873990.1
XM_017018502.1 Intron XP_016873991.1
XM_017018503.1 Intron XP_016873992.1
XM_017018504.1 Intron XP_016873993.1
XM_017018505.1 Intron XP_016873994.1
XM_017018506.1 Intron XP_016873995.1
XM_017018507.1 Intron XP_016873996.1
XM_017018508.1 Intron XP_016873997.1
XM_017018509.1 Intron XP_016873998.1
XM_017018510.1 Intron XP_016873999.1
Gene
NR1H3
Gene Name
nuclear receptor subfamily 1 group H member 3
There are no transcripts associated with this gene.

View Full Product Details