Product Details
- SNP ID
-
rs149697812
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:40981865 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCCAAGTGTGTTCTTTCTTGCTCT[A/G]GGGAGGGGGTGGGGCGGGGGAATGC
- Phenotype
-
MIM: 123860
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CTPS1
PubMed Links
Gene Details
- Gene
- CTPS1
- Gene Name
- CTP synthase 1
View Full Product Details