Product Details
- SNP ID
-
rs73645446
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:34992564 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGTTGGAAACAGCAGTTTCCAAAG[C/G]CTGTGAGTCACCCTGTGATCTGAGT
- Phenotype
-
MIM: 611328
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DNAJB5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79950507] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DNAJB5
- Gene Name
- DnaJ heat shock protein family (Hsp40) member B5
- Gene
- DNAJB5-AS1
- Gene Name
- DNAJB5 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.
View Full Product Details