Product Details

SNP ID
rs587674269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:112087725 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGCCGGGGTAACTGGCAGGTAA[C/T]TAGGCTTTGGAGTGGTATTTTAAAA
Phenotype
MIM: 607345
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM229B PubMed Links

Gene Details

Gene
FAM229B
Gene Name
family with sequence similarity 229 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033564.2 Intron NP_001028736.1
XM_017011174.1 Intron XP_016866663.1
XM_017011175.1 Intron XP_016866664.1
Gene
TUBE1
Gene Name
tubulin epsilon 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016262.4 Intron NP_057346.1
XM_011535875.2 Intron XP_011534177.1
XM_017010918.1 Intron XP_016866407.1

View Full Product Details