Product Details

SNP ID
rs147214081
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:114060441 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTGAGAAAACGTGGTGGGCTTT[C/T]TCTTGATTGGACTTGATCCCCACCC
Phenotype
MIM: 176797
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ZBTB16 PubMed Links

Gene Details

Gene
ZBTB16
Gene Name
zinc finger and BTB domain containing 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018011.1 Intron NP_001018011.1
NM_006006.4 Intron NP_005997.2
XM_005271658.4 Intron XP_005271715.1
XM_017018257.1 Intron XP_016873746.1
XM_017018258.1 Intron XP_016873747.1
XM_017018259.1 Intron XP_016873748.1

View Full Product Details