Product Details

SNP ID
hCV27834433
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:109440985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTCGGGCCCGACCTGAGCCACGC[A/G]GGCTTGGTGCCCACCTGTGCGCGCC
Phenotype
MIM: 607129 MIM: 603498
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MICAL1 PubMed Links

Gene Details

Gene
MICAL1
Gene Name
microtubule associated monooxygenase, calponin and LIM domain containing 1
There are no transcripts associated with this gene.

Gene
PPIL6
Gene Name
peptidylprolyl isomerase like 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111298.2 187 UTR 5 NP_001104768.2
NM_001286360.1 187 UTR 5 NP_001273289.1
NM_001286361.1 187 UTR 5 NP_001273290.1
NM_173672.4 187 UTR 5 NP_775943.1
Gene
SMPD2
Gene Name
sphingomyelin phosphodiesterase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003080.2 187 UTR 5 NP_003071.2

View Full Product Details