Product Details

SNP ID
rs148958953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:64388052 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGATTGGCTGGAATTACGGCCG[C/G]ATTCCGGAGTCCTTTCCAGCTCCCT
Phenotype
MIM: 172460
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MTHFD1 PubMed Links

Gene Details

Gene
MTHFD1
Gene Name
methylenetetrahydrofolate dehydrogenase, cyclohydrolase and formyltetrahydrofolate synthetase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005956.3 12 UTR 5 NP_005947.3

View Full Product Details