Product Details
- SNP ID
-
hCV27835943
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:107836935 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATAAGTTAAGGTAGGTTATTTTAGA[G/T]TACTAATGATTGGGTTTTTTTCTTT
- Phenotype
-
MIM: 300204
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MID2
PubMed Links
Gene Details
- Gene
- MID2
- Gene Name
- midline 2
View Full Product Details