Product Details

SNP ID
rs140711639
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34992519 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTGGTTGGGGGAAGGATAGTGT[C/T]GCCCCCCCTGCTAATGAGGGAGTTG
Phenotype
MIM: 611328
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJB5 PubMed Links

Gene Details

Gene
DNAJB5
Gene Name
DnaJ heat shock protein family (Hsp40) member B5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135004.2 Intron NP_001128476.2
NM_001135005.2 Intron NP_001128477.1
NM_012266.5 Intron NP_036398.3
XM_005251428.3 Intron XP_005251485.1
XM_006716751.2 Intron XP_006716814.1
XM_011517842.2 Intron XP_011516144.1
Gene
DNAJB5-AS1
Gene Name
DNAJB5 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
PHF24
Gene Name
PHD finger protein 24
There are no transcripts associated with this gene.

View Full Product Details