Product Details

SNP ID
rs114125269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:19479837 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGCCCAATCAGAAAGAAAGGAA[A/G]GCTGGGAACTAAGAAGCTATTGGTT
Phenotype
MIM: 603465 MIM: 601754
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDC45 PubMed Links

Gene Details

Gene
CDC45
Gene Name
cell division cycle 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178010.2 12 UTR 5 NP_001171481.1
NM_001178011.2 12 UTR 5 NP_001171482.1
NM_003504.4 12 UTR 5 NP_003495.1
XM_005261285.2 12 UTR 5 XP_005261342.1
XM_005261286.3 12 UTR 5 XP_005261343.1
XM_011530415.1 12 UTR 5 XP_011528717.1
XM_011530416.1 12 Intron XP_011528718.1
XM_011530417.2 12 UTR 5 XP_011528719.1
XM_011530418.2 12 UTR 5 XP_011528720.1
XM_017028966.1 12 Intron XP_016884455.1
XM_017028967.1 12 Intron XP_016884456.1
Gene
UFD1L
Gene Name
ubiquitin fusion degradation 1 like (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001035247.2 12 Intron NP_001030324.2
NM_005659.6 12 Intron NP_005650.2
XM_011530361.2 12 Intron XP_011528663.1

View Full Product Details