Product Details
- SNP ID
-
rs11048315
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:25874055 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACCTTAAGTAAACTCAGCCACCTC[A/G]CTGAGCCACACTGGTCCCTCCTGAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR4302
PubMed Links
Gene Details
- Gene
- MIR4302
- Gene Name
- microRNA 4302
There are no transcripts associated with this gene.
View Full Product Details