Product Details

SNP ID
rs190788605
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56116599 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCGGAAGTGACGACACCCGGCGC[G/T]CCATTAAATAGCCGTAGACGGAACT
Phenotype
MIM: 602145 MIM: 613315
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PA2G4 PubMed Links

Gene Details

Gene
PA2G4
Gene Name
proliferation-associated 2G4
There are no transcripts associated with this gene.

Gene
RPL41
Gene Name
ribosomal protein L41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001035267.1 Intron NP_001030344.1
NM_021104.1 Intron NP_066927.1
Gene
ZC3H10
Gene Name
zinc finger CCCH-type containing 10
There are no transcripts associated with this gene.

View Full Product Details