Product Details

SNP ID
rs114340939
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:118135997 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGACCCAGGAGCGCGAGAGGCCC[C/T]GCTCTGCCAGCTTCGGCCAATCAGA
Phenotype
MIM: 604591
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PEBP1 PubMed Links

Gene Details

Gene
PEBP1
Gene Name
phosphatidylethanolamine binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002567.3 114 UTR 5 NP_002558.1

View Full Product Details