Product Details
- SNP ID
-
hCV27845578
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:52340247 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACAAGTGAGAGCGCTCCACCCCGC[A/T]GTCCCCCCCGCCTCTCCTCCCTGGG
- Phenotype
-
MIM: 120470
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DCC
PubMed Links
Gene Details
- Gene
- DCC
- Gene Name
- DCC netrin 1 receptor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005215.3 |
76 |
UTR 5 |
|
|
NP_005206.2 |
View Full Product Details