Product Details

SNP ID
hCV27845872
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6931848 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAATCTAATTGTAATTCAATTAC[A/G]AAGTTTTTTTAAATCAAAGGACTCT
Phenotype
MIM: 605016
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2D3 PubMed Links

Gene Details

Gene
OR2D3
Gene Name
olfactory receptor family 2 subfamily D member 3
There are no transcripts associated with this gene.

Gene
ZNF215
Gene Name
zinc finger protein 215
There are no transcripts associated with this gene.

View Full Product Details