Product Details

SNP ID
rs41303051
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:211382789 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGGACATTTCTAGTCGACTGTTG[C/G]GCGTGCGCGCGCTGTGACGTTCCCT
Phenotype
MIM: 602356
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC00467 PubMed Links

Gene Details

Gene
LINC00467
Gene Name
long intergenic non-protein coding RNA 467
There are no transcripts associated with this gene.

Gene
TRAF5
Gene Name
TNF receptor associated factor 5
There are no transcripts associated with this gene.

View Full Product Details