Product Details

SNP ID
rs12073770
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:31410412 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAGCATGAGGCTGAGAGAGGAG[A/G]AGTCACCCGGACCCAGCCACACAGT
Phenotype
MIM: 614549
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SERINC2 PubMed Links

Gene Details

Gene
SERINC2
Gene Name
serine incorporator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199037.1 239 Missense Mutation AAG,GAG K10E NP_001185966.1
NM_001199038.1 239 Missense Mutation AAG,GAG K15E NP_001185967.1
NM_001199039.1 239 Intron NP_001185968.1
NM_018565.3 239 Intron NP_061035.2
NM_178865.4 239 Intron NP_849196.2

View Full Product Details