Product Details

SNP ID
rs928533465
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:49624752 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAATTTTGCCAATTCCACTAGAAT[C/G]TTCTATTTTTCATTAATGTCAATTC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DTWD1 PubMed Links

Gene Details

Gene
DTWD1
Gene Name
DTW domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144955.1 Intron NP_001138427.1
NM_020234.5 Intron NP_064619.2
Gene
FAM227B
Gene Name
family with sequence similarity 227 member B
There are no transcripts associated with this gene.

View Full Product Details