Product Details
- SNP ID
-
rs12480322
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:31848838 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATGATGAGCTGCTCCTTGGGGTGCT[C/G]TGTGGGGCCCGGGTCCTCCTCACCG
- Phenotype
-
MIM: 616776
MIM: 602939
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DUSP15
PubMed Links
Gene Details
- Gene
- DUSP15
- Gene Name
- dual specificity phosphatase 15
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001012644.2 |
877 |
Intron |
|
|
NP_001012662.1 |
NM_001320478.1 |
877 |
Intron |
|
|
NP_001307407.1 |
NM_001320479.1 |
877 |
Missense Mutation |
CAG,GAG |
Q232E |
NP_001307408.1 |
NM_080611.4 |
877 |
Intron |
|
|
NP_542178.2 |
NM_177991.2 |
877 |
Intron |
|
|
NP_817130.1 |
XM_017027654.1 |
877 |
Missense Mutation |
CAG,GAG |
Q272E |
XP_016883143.1 |
XM_017027655.1 |
877 |
Missense Mutation |
CAG,GAG |
Q269E |
XP_016883144.1 |
XM_017027656.1 |
877 |
Missense Mutation |
CAG,GAG |
Q272E |
XP_016883145.1 |
XM_017027657.1 |
877 |
Missense Mutation |
CAG,GAG |
Q235E |
XP_016883146.1 |
XM_017027658.1 |
877 |
Missense Mutation |
CAG,GAG |
Q169E |
XP_016883147.1 |
XM_017027659.1 |
877 |
Missense Mutation |
CAG,GAG |
Q169E |
XP_016883148.1 |
XM_017027660.1 |
877 |
Missense Mutation |
CAG,GAG |
Q169E |
XP_016883149.1 |
XM_017027661.1 |
877 |
Missense Mutation |
CAG,GAG |
Q169E |
XP_016883150.1 |
XM_017027662.1 |
877 |
Missense Mutation |
CAG,GAG |
Q169E |
XP_016883151.1 |
- Gene
- FOXS1
- Gene Name
- forkhead box S1
There are no transcripts associated with this gene.
View Full Product Details