Product Details

SNP ID
rs139618773
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:45176288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTTGAAGAACCCCTTAGAGGGAG[A/G]AAGAGGGGGCAGTTCTAATAGTACC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC01121 PubMed Links

Gene Details

Gene
LINC01121
Gene Name
long intergenic non-protein coding RNA 1121
There are no transcripts associated with this gene.

View Full Product Details