Product Details

SNP ID
rs121908318
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:142742343 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCACTCACCACAGCCCATGCTCC[A/G]GGCTGCCACGAGCAGCAGCTGCACA
Phenotype
MIM: 606119
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LYPD2 PubMed Links

Gene Details

Gene
LYPD2
Gene Name
LY6/PLAUR domain containing 2
There are no transcripts associated with this gene.

Gene
SLURP1
Gene Name
secreted LY6/PLAUR domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020427.2 69 Missense Mutation CGG,TGG R15W NP_065160.1
Gene
THEM6
Gene Name
thioesterase superfamily member 6
There are no transcripts associated with this gene.

View Full Product Details