Product Details

SNP ID
hCV27862369
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12892125 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGTCCCTTTGCAGTTTTTCATC[G/T]GGAGATCATTTCGGAGATGGGGGAG
Phenotype
MIM: 608801 MIM: 600599 MIM: 611487
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GCDH PubMed Links

Gene Details

Gene
GCDH
Gene Name
glutaryl-CoA dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000159.3 389 Missense Mutation CGG,CTG R94L NP_000150.1
NM_013976.3 389 Missense Mutation CGG,CTG R94L NP_039663.1
Gene
KLF1
Gene Name
Kruppel like factor 1
There are no transcripts associated with this gene.

Gene
SYCE2
Gene Name
synaptonemal complex central element protein 2
There are no transcripts associated with this gene.

View Full Product Details