Product Details

SNP ID
rs4455975
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:126620920 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGGGTAGGTGGAGGAGAGGTCTG[A/G]GGGATTTGTCTATTTCTCAGAAGGA
Phenotype
MIM: 602575
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LMX1B PubMed Links

Gene Details

Gene
LMX1B
Gene Name
LIM homeobox transcription factor 1 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174146.1 Intron NP_001167617.1
NM_001174147.1 Intron NP_001167618.1
NM_002316.3 Intron NP_002307.2

View Full Product Details