Product Details
- SNP ID
-
rs4842071
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:136010451 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTTCACACGTACTAGTCGCCCACCC[A/G]GGACCCCCTTTTTCCCCCAGTGGGA
- Phenotype
-
MIM: 615786
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NACC2
PubMed Links
Gene Details
- Gene
- NACC2
- Gene Name
- NACC family member 2
View Full Product Details