Product Details

SNP ID
rs4958179
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:134312871 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTCAATAACCCATTATTAAATCA[C/G]CTCTTTTCTGTTTCCCTTTCTTGGT
Phenotype
MIM: 608459
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDKL3 PubMed Links

Gene Details

Gene
CDKL3
Gene Name
cyclin dependent kinase like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113575.1 Intron NP_001107047.1
NM_001300853.1 Intron NP_001287782.1
NM_016508.3 Intron NP_057592.2

View Full Product Details