Product Details

SNP ID
rs4903522
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:76963725 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGAGGCAAGACTGGACAAAGAGA[C/G]ACCCGTCAAGAGTTTGTTACAGGCC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC01629 PubMed Links

Gene Details

Gene
LINC01629
Gene Name
long intergenic non-protein coding RNA 1629
There are no transcripts associated with this gene.

View Full Product Details