Product Details

SNP ID
rs4634866
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:35179819 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAAATAGACCTGAGTTTTTATTT[C/T]TAACACCCAATCACTAAACCACGGC
Phenotype
MIM: 605199
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SFPQ PubMed Links

Gene Details

Gene
SFPQ
Gene Name
splicing factor proline and glutamine rich
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005066.2 Intron NP_005057.1

View Full Product Details