Product Details

SNP ID
rs4723713
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:37848752 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGTATAGCTGCTGTCTGGAGCAGG[A/G]GCCTCGAGACGCCGCTCAAGTCTCA
Phenotype
MIM: 607421
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
NME8 PubMed Links
Additional Information
For this assay, SNP(s) [rs80144288] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
NME8
Gene Name
NME/NM23 family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016616.4 Intron NP_057700.3

View Full Product Details