Product Details

SNP ID
rs4852325
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:73986488 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCAGCGCCAGGTGATGGTAGGG[A/G]GCTTCCCGGGGTCTGGGCTCTCCAT
Phenotype
MIM: 613555
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DGUOK-AS1 PubMed Links

Gene Details

Gene
DGUOK-AS1
Gene Name
DGUOK antisense RNA 1
There are no transcripts associated with this gene.

Gene
TET3
Gene Name
tet methylcytosine dioxygenase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287491.1 85 Missense Mutation AGC,GGC S29G NP_001274420.1

View Full Product Details