Product Details
- SNP ID
-
rs4891246
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:75216889 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGGCATGGTCCACTGGGCCGGGAC[C/G]CACTCCTGTTTCTGCCTGTGCATTT
- Phenotype
-
MIM: 614427
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TSHZ1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74540049] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TSHZ1
- Gene Name
- teashirt zinc finger homeobox 1
- Gene
- ZADH2
- Gene Name
- zinc binding alcohol dehydrogenase domain containing 2
There are no transcripts associated with this gene.
View Full Product Details