Product Details

SNP ID
rs4945817
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:108752108 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGATCTCCCCCCATGAAATGAAGGA[A/G]AATATGCTAAACTAGTGGCTGCTGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00222 PubMed Links

Gene Details

Gene
LINC00222
Gene Name
long intergenic non-protein coding RNA 222
There are no transcripts associated with this gene.

View Full Product Details