Product Details

SNP ID
rs4791840
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7840875 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATTTGCTGCACAGCCTCCTACTTT[A/C]TCCTCTAGAGGACGTGAGACATTGG
Phenotype
MIM: 603333 MIM: 611577
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DNAH2 PubMed Links

Gene Details

Gene
DNAH2
Gene Name
dynein axonemal heavy chain 2
There are no transcripts associated with this gene.

Gene
KDM6B
Gene Name
lysine demethylase 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080424.1 Intron NP_001073893.1
XM_005256549.3 Intron XP_005256606.1
XM_005256550.4 Intron XP_005256607.1
XM_005256551.3 Intron XP_005256608.1
XM_005256552.4 Intron XP_005256609.1
XM_006721483.3 Intron XP_006721546.1
XM_011523750.2 Intron XP_011522052.1
XM_011523752.2 Intron XP_011522054.1
XM_017024381.1 Intron XP_016879870.1

View Full Product Details