Product Details

SNP ID
rs4806887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2902421 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCCGTCAGGATAACACCTGTTTAT[C/T]GTGGGGTACAGGCTAAGCATGGCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF57 PubMed Links

Gene Details

Gene
ZNF57
Gene Name
zinc finger protein 57
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319083.1 Intron NP_001306012.1
NM_173480.2 Intron NP_775751.1
XM_011527682.2 Intron XP_011525984.1

View Full Product Details